COM448687281 Combi-Blocks, Inc.

methyl 3-(4-chloro-2-methylphenyl)benzoate

  • MDL number MFCD27578830
  • Cas Number : 1820739-77-2
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2443
Availability details for
Quantity
  • Availability
  • Add to Cart