COM448687284 Combi-Blocks, Inc.

methyl 4-(3,5-dimethoxyphenyl)-2-fluorobenzoate

  • MDL number MFCD27578833
  • Cas Number : 1820711-12-3
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2447
Availability details for
Quantity
  • Availability
  • Add to Cart