COM448687288 Combi-Blocks, Inc.

methyl 4-(2-chlorophenyl)-2-fluorobenzoate

  • MDL number MFCD27578835
  • Cas Number : 1820603-44-8
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2455
Availability details for
Quantity
  • Availability
  • Add to Cart