COM448687290 Combi-Blocks, Inc.

methyl 2-fluoro-4-(4-fluorophenyl)benzoate

  • MDL number MFCD27578837
  • Cas Number : 1098595-16-4
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2457
Availability details for
Quantity
  • Availability
  • Add to Cart