COM448687292 Combi-Blocks, Inc.

methyl 4-(3-fluoro-5-nitrophenyl)benzoate

  • MDL number MFCD27578839
  • Cas Number : 1820641-03-9
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2459
Availability details for
Quantity
  • Availability
  • Add to Cart