COM448687294 Combi-Blocks, Inc.

Methyl 5-(4-(trifluoromethyl)phenyl)pyridine-3-carboxylate

  • MDL number MFCD06802192
  • Cas Number : 893734-81-1
  • Molecular Weight : 281.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2462
Availability details for
Quantity
  • Availability
  • Add to Cart