COM448687297 Combi-Blocks, Inc.

(4-Chloro-phenylamino)-acetic acid methyl ester

  • MDL number MFCD00461439
  • Cas Number : 131770-31-5
  • Molecular Weight : 199.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2465
Availability details for
Quantity
  • Availability
  • Add to Cart