COM448687298 Combi-Blocks, Inc.

methyl 2-{3-[4-(methylsulfanyl)phenyl]phenyl}acetate

  • MDL number MFCD27578842
  • Cas Number : 1820650-85-8
  • Molecular Weight : 272.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2466
Availability details for
Quantity
  • Availability
  • Add to Cart