COM448687302 Combi-Blocks, Inc.

methyl 3-(2-ethylphenyl)benzoate

  • MDL number MFCD05979028
  • Cas Number : 1820619-10-0
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2473
Availability details for
Quantity
  • Availability
  • Add to Cart