COM448687306 Combi-Blocks, Inc.

Methyl 2-chloro-5-phenylnicotinate

  • MDL number MFCD18256773
  • Cas Number : 10177-09-0
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2479
Availability details for
Quantity
  • Availability
  • Add to Cart