COM448687308 Combi-Blocks, Inc.

methyl 4-(3-cyanophenyl)-3-fluorobenzoate

  • MDL number MFCD27578848
  • Cas Number : 1820642-19-0
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2481
Availability details for
Quantity
  • Availability
  • Add to Cart