COM448687312 Combi-Blocks, Inc.

Methyl 3-(4-nitrobenzenesulfonamido)propanoate

  • MDL number MFCD00553482
  • Cas Number : 339191-82-1
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2485
Availability details for
Quantity
  • Availability
  • Add to Cart