COM448687313 Combi-Blocks, Inc.

methyl 3-methoxy-5-(4-methylphenyl)benzoate

  • MDL number MFCD27578851
  • Cas Number : 1820638-58-1
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2486

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2486
Availability details for
Quantity
  • Availability
  • Add to Cart