COM448687317 Combi-Blocks, Inc.

methyl 2-(3,4-dimethoxyphenyl)benzoate

  • MDL number MFCD27578855
  • Cas Number : 34169-91-0
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2492
Availability details for
Quantity
  • Availability
  • Add to Cart