COM448687318 Combi-Blocks, Inc.

Methyl 3-chloro-2'-methyl-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD23134134
  • Cas Number : 1238634-35-9
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2493
Availability details for
Quantity
  • Availability
  • Add to Cart