COM448687320 Combi-Blocks, Inc.

methyl 4-(3-chloro-4-methylphenyl)-2-fluorobenzoate

  • MDL number MFCD27578857
  • Cas Number : 1820707-70-7
  • Molecular Weight : 278.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2495
Availability details for
Quantity
  • Availability
  • Add to Cart