COM448687321 Combi-Blocks, Inc.

methyl 4-(4-fluoro-3-methoxyphenyl)-3-methylbenzoate

  • MDL number MFCD27578858
  • Cas Number : 1820649-43-1
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2496
Availability details for
Quantity
  • Availability
  • Add to Cart