COM448687322 Combi-Blocks, Inc.

[3-(4-methoxy-4-oxobutanamido)phenyl]boronic acid

  • MDL number MFCD27578859
  • Molecular Weight : 251.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2498
Availability details for
Quantity
  • Availability
  • Add to Cart