COM448687329 Combi-Blocks, Inc.

Methyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate

  • MDL number MFCD20922247
  • Cas Number : 79247-82-8
  • Molecular Weight : 290.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2510
Availability details for
Quantity
  • Availability
  • Add to Cart