COM448687330 Combi-Blocks, Inc.

methyl 3-(4-chlorophenyl)-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578865
  • Cas Number : 1820710-89-1
  • Molecular Weight : 314.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2511
Availability details for
Quantity
  • Availability
  • Add to Cart