COM448687331 Combi-Blocks, Inc.

methyl 6-(2,4-difluorophenyl)pyridine-3-carboxylate

  • MDL number MFCD27578866
  • Cas Number : 938433-58-0
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2515
Availability details for
Quantity
  • Availability
  • Add to Cart