COM448687337 Combi-Blocks, Inc.

methyl 3-[3-(ethylcarbamoyl)phenyl]-5-methoxybenzoate

  • MDL number MFCD27578871
  • Cas Number : 1820686-37-0
  • Molecular Weight : 313.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2522
Availability details for
Quantity
  • Availability
  • Add to Cart