COM448687339 Combi-Blocks, Inc.

4-Phenylpyridine-3-carboxylic acid methyl ester

  • MDL number MFCD18417739
  • Cas Number : 70647-05-1
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2524
Availability details for
Quantity
  • Availability
  • Add to Cart