COM448687345 Combi-Blocks, Inc.

methyl 6-[4-(benzyloxy)phenyl]pyridine-3-carboxylate

  • MDL number MFCD27578875
  • Cas Number : 1820711-30-5
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2533
Availability details for
Quantity
  • Availability
  • Add to Cart