COM448687347 Combi-Blocks, Inc.

Methyl imidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD12912213
  • Cas Number : 1220397-13-6
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2535
Availability details for
Quantity
  • Availability
  • Add to Cart