COM448687351 Combi-Blocks, Inc.

methyl 3-(3,5-dichlorophenyl)-5-methoxybenzoate

  • MDL number MFCD27578878
  • Cas Number : 1820739-86-3
  • Molecular Weight : 311.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2540

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2540
Availability details for
Quantity
  • Availability
  • Add to Cart