COM448687352 Combi-Blocks, Inc.

Methyl 2-[2-(2-aminoacetamido)acetamido]acetate

  • MDL number MFCD16042797
  • Cas Number : 31681-05-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2541
Availability details for
Quantity
  • Availability
  • Add to Cart