COM448687362 Combi-Blocks, Inc.

1-benzyl 2-methyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD27578886
  • Cas Number : 690211-34-8
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2555
Availability details for
Quantity
  • Availability
  • Add to Cart