COM448687363 Combi-Blocks, Inc.

2-Fluoro-3'-(trifluoromethoxy)biphenyl-4-carboxylic acid methyl ester

  • MDL number MFCD18408087
  • Cas Number : 1261616-51-6
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2557
Availability details for
Quantity
  • Availability
  • Add to Cart