COM448687364 Combi-Blocks, Inc.

methyl 4-(3,4-difluorophenyl)benzoate

  • MDL number MFCD27578887
  • Cas Number : 1382448-99-8
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2558
Availability details for
Quantity
  • Availability
  • Add to Cart