COM448687365 Combi-Blocks, Inc.

methyl 4-chloro-3-(morpholine-4-sulfonyl)benzoate

  • MDL number MFCD01203069
  • Cas Number : 352668-16-7
  • Molecular Weight : 319.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2560
Availability details for
Quantity
  • Availability
  • Add to Cart