COM448687371 Combi-Blocks, Inc.

methyl 3-(3,5-difluorophenyl)-5-methoxybenzoate

  • MDL number MFCD27578890
  • Cas Number : 1820747-70-3
  • Molecular Weight : 278.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2568
Availability details for
Quantity
  • Availability
  • Add to Cart