COM448687372 Combi-Blocks, Inc.

4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid methyl ester

  • MDL number MFCD14698096
  • Cas Number : 786-52-7
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2569
Availability details for
Quantity
  • Availability
  • Add to Cart