COM448687373 Combi-Blocks, Inc.

methyl 3-(4-bromophenyl)-2-acetamidopropanoate

  • MDL number MFCD27578891
  • Cas Number : 151073-59-5
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2572
Availability details for
Quantity
  • Availability
  • Add to Cart