COM448687375 Combi-Blocks, Inc.

methyl 2-(4-tert-butylphenyl)pyridine-4-carboxylate

  • MDL number MFCD27578893
  • Cas Number : 1251898-44-8
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2575
Availability details for
Quantity
  • Availability
  • Add to Cart