COM448687379 Combi-Blocks, Inc.

hydrate methyl 6-hydroxypyridazine-3-carboxylate

  • MDL number MFCD27578895
  • Cas Number : 1820650-57-4
  • Molecular Weight : 172.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2580
Availability details for
Quantity
  • Availability
  • Add to Cart