COM448687381 Combi-Blocks, Inc.

3-(4-Chloro-phenyl)-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD20441176
  • Cas Number : 347174-25-8
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2584
Availability details for
Quantity
  • Availability
  • Add to Cart