COM448687383 Combi-Blocks, Inc.

Methyl 3-(2-thienyl)benzoate

  • MDL number MFCD06204245
  • Cas Number : 17595-85-6
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2586
Availability details for
Quantity
  • Availability
  • Add to Cart