COM448687386 Combi-Blocks, Inc.

P-Trifluoroacetylbenzoic acid methyl ester

  • MDL number MFCD25965464
  • Cas Number : 62263-12-1
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2589
Availability details for
Quantity
  • Availability
  • Add to Cart