COM448687387 Combi-Blocks, Inc.

(3-Fluoro-benzoylamino)-acetic acid methyl ester

  • MDL number MFCD03992420
  • Cas Number : 131648-59-4
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2594

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2594
Availability details for
Quantity
  • Availability
  • Add to Cart