COM448687390 Combi-Blocks, Inc.

methyl 3-(3-fluorophenyl)-5-methoxybenzoate

  • MDL number MFCD27578900
  • Cas Number : 1427407-27-9
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2597
Availability details for
Quantity
  • Availability
  • Add to Cart