COM448687391 Combi-Blocks, Inc.

methyl 3-(2H-1,3-benzodioxol-5-yl)-5-methoxybenzoate

  • MDL number MFCD27578901
  • Cas Number : 1820618-35-6
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2598
Availability details for
Quantity
  • Availability
  • Add to Cart