COM448687394 Combi-Blocks, Inc.

methyl 3-[4-(hydroxymethyl)phenyl]pyridine-2-carboxylate

  • MDL number MFCD27578903
  • Cas Number : 1808096-72-1
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2603
Availability details for
Quantity
  • Availability
  • Add to Cart