COM448687395 Combi-Blocks, Inc.

methyl 3-(6-chloro-2-methylpyridin-3-yl)benzoate

  • MDL number MFCD27578904
  • Cas Number : 1820673-37-7
  • Molecular Weight : 261.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2604
Availability details for
Quantity
  • Availability
  • Add to Cart