COM448687396 Combi-Blocks, Inc.

methyl 2-fluoro-4-(4-methoxyphenyl)benzoate

  • MDL number MFCD27578905
  • Cas Number : 454465-73-7
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2605
Availability details for
Quantity
  • Availability
  • Add to Cart