COM448687398 Combi-Blocks, Inc.

methyl 3-fluoro-4-[2-(methylsulfanyl)phenyl]benzoate

  • MDL number MFCD27578907
  • Cas Number : 1820674-24-5
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2607
Availability details for
Quantity
  • Availability
  • Add to Cart