COM448687399 Combi-Blocks, Inc.

methyl (2S)-3-carbamoyl-2-(1,3-dioxoisoindol-2-yl)propanoate

  • MDL number MFCD27578908
  • Cas Number : 1820574-75-1
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2608

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2608
Availability details for
Quantity
  • Availability
  • Add to Cart