COM448687400 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-cyanophenyl)propanoate

  • MDL number MFCD27578909
  • Cas Number : 298693-79-5
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2609
Availability details for
Quantity
  • Availability
  • Add to Cart