COM448687402 Combi-Blocks, Inc.

methyl (2S)-2-amino-4-(methylselanyl)butanoate

  • MDL number MFCD27578911
  • Cas Number : 821766-18-1
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2612
Availability details for
Quantity
  • Availability
  • Add to Cart