COM448687403 Combi-Blocks, Inc.

methyl 3-(3-chloro-5-fluorophenyl)-5-nitrobenzoate

  • MDL number MFCD27578912
  • Cas Number : 1820706-77-1
  • Molecular Weight : 309.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2613
Availability details for
Quantity
  • Availability
  • Add to Cart